accessibility menu, dialog, popup

UserName

CAS RN 138112-76-2

H3CONHCH3O

CAS RN 138112-76-2

IUPAC Name: N-[2-(7-methoxynaphthalen-1-yl)ethyl]acetamide
Synonyms: agomelatine
Molecular Weight (g/mol): 243.31
Molecular Formula: C15H17NO2
InChi Key: YJYPHIXNFHFHND-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC=C(CCNC(C)=O)C2=C1
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results