missing translation for 'onlineSavingsMsg'
Learn More

Agomelatine, Mikromol™

Product Code. 17933056
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933056 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17933056 Supplier Mikromol Supplier No. MM3583.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Agomelatine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Agomelatine
CAS 138112-76-2
Analyte or Component Names Agomelatine
Shipping Condition Room Temperature
Molecular Formula C15 H17 N O2
Certifications/Compliance ISO 17034
Synonym N-[2-(7-Methoxy-1-naphthalenyl)ethyl]-acetamide, AGO 178, Agomelatine, N-[2-(7-Methoxy-1-naphthyl)ethyl]acetamide, N-[2-(7-Methoxynaphthalen-1-yl)ethyl]acetamide, S 20098, Valdoxan
InChI Formula InChI=1S/C15H17NO2/c1-11(17)16-9-8-13-5-3-4-12-6-7-14(18-2)10-15(12)13/h3-7,10H,8-9H2,1-2H3,(H,16,17)
SMILES COc1ccc2cccc(CCNC(=O)C)c2c1
IUPAC Name N-[2-(7-methoxynaphthalen-1-yl)ethyl]acetamide
Molecular Weight (g/mol) 243.3
Quantity 250 mg
Formula Weight 243.1259 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.