missing translation for 'onlineSavingsMsg'
Learn More

Agomelatine, TRC

Product Code. 17942875
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942875 5 mg 5mg
17920765 10 mg 10mg
17942335 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17942875 Supplier Toronto Research Chemicals Supplier No. TRCA4300005MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138112-76-2
Molecular Formula C15 H17 N O2
Molecular Weight (g/mol) 243.3
Synonym N-[2-(7-Methoxy-1-naphthalenyl)ethyl]-acetamide, AGO 178, Agomelatine, N-[2-(7-Methoxy-1-naphthyl)ethyl]acetamide, N-[2-(7-Methoxynaphthalen-1-yl)ethyl]acetamide, S 20098, Valdoxan
IUPAC Name N-[2-(7-methoxynaphthalen-1-yl)ethyl]acetamide
SMILES COc1ccc2cccc(CCNC(=O)C)c2c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H17NO2/c1-11(17)16-9-8-13-5-3-4-12-6-7-14(18-2)10-15(12)13/h3-7,10H,8-9H2,1-2H3,(H,16,17)
Formula Weight 243.1259
Percent Purity >95%
Chemical Name or Material Agomelatine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.