CAS RN 1329834-57-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Romifidine-d4 Hydrochloride, TRC
CAS: 1329834-57-2 Molecular Formula: C9H6D4BrClFN3 Molecular Weight (g/mol): 298.58 Synonym: N-(2-Bromo-6-fluorophenyl)-4,5-dihydro-1H-imidazol-2-amine Hydrochloride,STH 2130Cl,Sedivet SMILES: [2H]C1(C(N=C(N1)NC2=C(C=CC=C2Br)F)([2H])[2H])[2H].Cl