Learn More
Romifidine-d4 Hydrochloride, TRC
Product Code. 30424400
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424400 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30424400 Supplier Toronto Research Chemicals Supplier No. TRCR64005210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1329834-57-2
Molecular Formula C9H6D4BrClFN3
Molecular Weight (g/mol) 298.58
Synonym N-(2-Bromo-6-fluorophenyl)-4,5-dihydro-1H-imidazol-2-amine Hydrochloride, STH 2130Cl, Sedivet
SMILES [2H]C1(C(N=C(N1)NC2=C(C=CC=C2Br)F)([2H])[2H])[2H].Cl
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C9H9BrFN3.ClH/c10-6-2-1-3-7(11)8(6)14-9-12-4-5-13-9;/h1-3H,4-5H2,(H2,12,13,14);1H/i4D2,5D2;
Formula Weight 296.9982
Chemical Name or Material Romifidine-d4 Hydrochloride