accessibility menu, dialog, popup

UserName

OOOYY

CAS RN 1314-36-9

IUPAC Name: diyttrium(3+) trioxidandiide
Synonyms: yttrium(III) oxide
Molecular Weight (g/mol): 225.81
Molecular Formula: O3Y2
InChi Key: RUDFQVOCFDJEEF-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Y+3].[Y+3]
Quantity 
  • (4)
  • (3)
  • (5)
  • (1)
  • (5)
  • (4)
  • (5)
  • (4)
Physical Form 
  • (3)
  • (25)
  • (1)
Percent Purity 
  • (8)
  • (5)
  • (6)
  • (3)
Grade 
  • (2)
  • (6)
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (6)
  • (27)
  • (1)
  • (4)
  • (3)
  • (5)
  • (1)
  • (5)
  • (4)
  • (5)
  • (4)
  • (3)
  • (25)
  • (1)
  • (8)
  • (5)
  • (6)
  • (3)
  • (2)
  • (6)
  • (3)
  • (6)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)