missing translation for 'onlineSavingsMsg'
Learn More

Yttrium Oxide, 99%+, MilliporeSigma™

Product Code. 15885640
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15885640 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15885640 Supplier Supelco Supplier No. 1124120025
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity EMSURE™ grade yttrium oxide for analytical applications.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 2410°C
CAS 1314-36-9
Color Light Gray
Physical Form Solid
Quantity 25 g
Assay Percent Range 0.99%
Molecular Formula O3Y2
MDL Number MFCD00011473
Synonym Yttrium (III) Oxide 99
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Y+3].[Y+3]
IUPAC Name Diyttrium(3+) Trioxidandiide
Molecular Weight (g/mol) 225.81
Formula Weight 225.81g/moL
Odor Odorless
Packaging Plastic Bottle
Product Line EMSURE
Appearance Light Gray Solid
Specific Gravity 5.0
Density 5.0 g/cm³
Chemical Name or Material Yttrium Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.