accessibility menu, dialog, popup

UserName

NaNaNH2NON(R)(S)O(R)OPHOOOO

CAS RN 13085-50-2

IUPAC Name: disodium (2R,3S,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-2-[(hydrogen phosphonatooxy)methyl]oxolan-3-olate
Synonyms: disodium (2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-2-[(hydrogen phosphonatooxy)methyl]oxolan-3-olate
Molecular Weight (g/mol): 351.16
Molecular Formula: C9H12N3Na2O7P
InChi Key: HKNBOJGORNDJOL-CDNBRZBRSA-M
SMILES: [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@H]1C[C@H]([O-])[C@@H](COP(O)([O-])=O)O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results