Learn More
2'-Deoxycytidine-5'-monophosphate disodium salt, Thermo Scientific Chemicals
Product Code. 15427049
Change view
Click to view available options
Quantity:
25 mg
1 g
Unit Size:
1g
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427049 1 g 1g
15437049 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15427049 Supplier Thermo Scientific Alfa Aesar Supplier No. J65724.03
  • ONLINE EXCLUSIVE PRICE
    92.60 EUR
    83.65 EUR / 1g
    Save 8.95 EUR
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
2?-Deoxycytidine 5-monophosphate (dCMP) is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′-Deoxycytidine 5-monophosphate (dCMP) is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.
Solubility
Soluble in 200 mg plus 4 mL water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13085-50-2
Molecular Formula C9H14N3Na2O7P
Molecular Weight (g/mol) 353.178
MDL Number MFCD00069774
InChI Key ACIYKIIBZKDNAD-CDNBRZBRSA-N
Synonym 2'-deoxycytidine-5'-monophosphate disodium salt
PubChem CID 131846647
IUPAC Name [(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O.[Na].[Na]
Specifications
Color White
Quantity 1 g
Solubility Information Soluble in 200 mg plus 4 mL water.
Formula Weight 351.16
Physical Form Powder
Chemical Name or Material 2'-Deoxycytidine-5'-monophosphate disodium salt