CAS RN 12678-07-8
IUPAC Name:
6-[(2-{[2-carboxylato-6-({2-[(2-carboxylato-4,5,6-trihydroxyoxan-3-yl)oxy]-3-acetamido-5-hydroxy-6-[(sulfonatooxy)methyl]oxan-4-yl}oxy)-4,5-dihydroxyoxan-3-yl]oxy}-3-acetamido-5-hydroxy-6-[(sulfonatooxy)methyl]oxan-4-yl)oxy]-3-({3-acetamido-4,5-dihydroxy-6-[(sulfonatooxy)methyl]oxan-2-yl}oxy)-4,5-dihydroxyoxane-2-carboxylate
Synonyms:
6-[(2-{[2-carboxylato-6-({2-[(2-carboxylato-4,5,6-trihydroxyoxan-3-yl)oxy]-3-acetamido-5-hydroxy-6-[(sulfonatooxy)methyl]oxan-4-yl}oxy)-4,5-dihydroxyoxan-3-yl]oxy}-3-acetamido-5-hydroxy-6-[(sulfonatooxy)methyl]oxan-4-yl)oxy]-3-({3-acetamido-4,5-dihydroxy-6-[(sulfonatooxy)methyl]oxan-2-yl}oxy)-4,5-dihydroxyoxane-2-carboxylate
Molecular Weight (g/mol):
1390.1
Molecular Formula:
C42H59N3O43S3
InChi Key:
ABHQVWCLVQDEKE-UHFFFAOYNA-H
SMILES:
CC(=O)NC1C(O)C(O)C(COS([O-])(=O)=O)OC1OC1C(O)C(O)C(OC2C(O)C(COS([O-])(=O)=O)OC(OC3C(O)C(O)C(OC4C(O)C(COS([O-])(=O)=O)OC(OC5C(O)C(O)C(O)OC5C([O-])=O)C4NC(C)=O)OC3C([O-])=O)C2NC(C)=O)OC1C([O-])=O
1
–
2
of
2
results
1
Chondroitin Sulfate C Sodium Salt (Technical Grade), TRC
CAS: 12678-07-8 Molecular Formula: (C14 H19 N O14 S . 2 Na)n Molecular Weight (g/mol): 1390.1 Synonym: Chondoitrin 6-sulfate sodium salt,Sodium chondroitin sulfate C IUPAC Name: 3-[3-acetamido-4-[5-[3-acetamido-4-[5-[3-acetamido-4,5-dihydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-6-carboxylato-3,4-dihydroxyoxan-2-yl]oxy-5-hydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-6-carboxylato-3,4-dihydroxyoxan-2-yl]oxy-5-hydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-4,5,6-trihydroxyoxane-2-carboxylate SMILES: [Na+].[Na+].[*]O[C@@H]1C(O)C(O)[C@H](O[C@@H]2C(O)C(NC(=O)C)[C@H]([*])OC2COS(=O)(=O)[O-])OC1C(=O)[O-]