missing translation for 'onlineSavingsMsg'
Learn More

Chondroitin Sulfate C Sodium Salt (Technical Grade), TRC

Product Code. 17977254
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17977254 2.5 g 2.5g
17959914 25 g 25g
17918004 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17977254 Supplier Toronto Research Chemicals Supplier No. TRCC4333782.5G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12678-07-8
Molecular Formula (C14 H19 N O14 S . 2 Na)n
Molecular Weight (g/mol) 1390.1
Synonym Chondoitrin 6-sulfate sodium salt, Sodium chondroitin sulfate C
IUPAC Name 3-[3-acetamido-4-[5-[3-acetamido-4-[5-[3-acetamido-4,5-dihydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-6-carboxylato-3,4-dihydroxyoxan-2-yl]oxy-5-hydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-6-carboxylato-3,4-dihydroxyoxan-2-yl]oxy-5-hydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-4,5,6-trihydroxyoxane-2-carboxylate
SMILES [Na+].[Na+].[*]O[C@@H]1C(O)C(O)[C@H](O[C@@H]2C(O)C(NC(=O)C)[C@H]([*])OC2COS(=O)(=O)[O-])OC1C(=O)[O-]

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C42H65N3O43S3/c1-7(46)43-13-19(52)16(49)10(4-75-89(66,67)68)78-38(13)84-29-21(54)24(57)41(87-32(29)35(61)62)83-27-15(45-9(3)48)40(80-12(18(27)51)6-77-91(72,73)74)86-30-22(55)25(58)42(88-33(30)36(63)64)82-26-14(44-8(2)47)39(79-11(17(26)50)5-76-90(69,70)71)85-28-20(53)23(56)37(65)81-31(28)34(59)60/h10-33,37-42,49-58,65H,4-6H2,1-3H3,(H,43,46)(H,44,47)(H,45,48)(H,59,60)(H,61,62)(H,63,64)(H,66,67,68)(H,69,70,71)(H,72,73,74)/p-6
Formula Weight 499.0608
Chemical Name or Material Chondroitin sulfate C sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.