missing translation for 'onlineSavingsMsg'
Learn More

Chondroitin Sulfate C, MP Biomedicals

Product Code. 11404336 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404336 10 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11404336 Supplier MP Biomedicals Supplier No. 08970551

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

From Shark Cartilage Sodium Salt Inorganic sulfur and chloride are not detected. The Mrin 4-8 x 104and the 4 to 6 sulfate ratio is 10:90

TRUSTED_SUSTAINABILITY

Specifications

CAS 12678-07-8
Molecular Formula C42H59N3O43S3-6
Synonym yl-1->4 hexopyranuronate, chondroitin 6-sulfate sodium salt?, 2-acetamido-2-deoxy-6-o-sulfonatohexopyranosyl-1->4 hexopyranuronosyl-1->3-2-acetamido-2-deoxy-6-o-sulfonatohexopyranosyl-1->4 hexopyranuronosyl-1->3-2-acetamido-2-deoxy-6-o-sulfonatohexopyranos, 3-3-acetamido-4-5-3-acetamido-4-5-3-acetamido-4,5-dihydroxy-6-sulfonatooxymethyl-2-oxanyl oxy-6-carboxylato-3,4-dihydroxy-2-oxanyl oxy-5-hydroxy-6-sulfonatooxymethyl-2-oxanyl oxy-6-carboxylato-3,4-dihydroxy-2-oxanyl oxy-5-hydroxy-6-sulf, 3-3-acetamido-4-5-3-acetamido-4-5-3-acetamido-4,5-bis oxidanyl-6-sulfonatooxymethyl oxan-2-yl oxy-6-carboxylato-3,4-bis oxidanyl oxan-2-yl oxy-5-oxidanyl-6-sulfonatooxymethyl oxan-2-yl oxy-6-carboxylato-3,4-bis oxidanyl oxan-2-yl oxy-5-oxidanyl-6, 6-2-2-carboxylato-6-2-2-carboxylato-4,5,6-trihydroxyoxan-3-yl oxy-3-acetamido-5-hydroxy-6-sulfonatooxy methyl oxan-4-yl oxy-4,5-dihydroxyoxan-3-yl oxy-3-acetamido-5-hydroxy-6-sulfonatooxy methyl oxan-4-yl oxy-3-3-acetamido-4,5-dihydroxy-6-sulfonatooxy met
Quantity 10 mg
InChI Key ABHQVWCLVQDEKE-UHFFFAOYSA-H
SMILES CC(=O)NC1C(C(C(OC1OC2C(C(C(OC2C(=O)[O-])OC3C(C(OC(C3O)COS(=O)(=O)[O-])OC4C(C(C(OC4C(=O)[O-])OC5C(C(OC(C5O)COS(=O)(=O)[O-])OC6C(C(C(OC6C(=O)[O-])O)O)O)NC(=O)C)O)O)NC(=O)C)O)O)COS(=O)(=O)[O-])O)O
IUPAC Name 3-[3-acetamido-4-[5-[3-acetamido-4-[5-[3-acetamido-4,5-dihydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-6-carboxylato-3,4-dihydroxyoxan-2-yl]oxy-5-hydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-6-carboxylato-3,4-dihydroxyoxan-2-yl]oxy-5-hydroxy-6-(sulfonatooxym
Molecular Weight (g/mol) 1390.092
PubChem CID 42609007
Chemical Name or Material Chondroitin Sulfate C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.