accessibility menu, dialog, popup

UserName

CH3OONNNNOOH3COOCH3OOCH3

CAS RN 126320-57-8

IUPAC Name: ethyl 2-[4,8,11-tris(2-ethoxy-2-oxoethyl)-1,4,8,11-tetraazacyclotetradecan-1-yl]acetate
Synonyms: ethyl 2-[4,8,11-tris(2-ethoxy-2-oxoethyl)-1,4,8,11-tetraazacyclotetradecan-1-yl]acetate
Molecular Weight (g/mol): 544.69
Molecular Formula: C26H48N4O8
InChi Key: HGPDBLIYOCNCEH-UHFFFAOYSA-N
SMILES: CCOC(=O)CN1CCCN(CC(=O)OCC)CCN(CC(=O)OCC)CCCN(CC(=O)OCC)CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results