missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,4,8,11-Tetrakis(ethoxycarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane

Product Code. 15467615
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467615 250 mg 250mg
11356819 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467615 Supplier Alfa Aesar™ Supplier No. H26755.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 126320-57-8
Molecular Formula C26H48N4O8
Molecular Weight (g/mol) 544.69
MDL Number MFCD09263316
InChI Key HGPDBLIYOCNCEH-UHFFFAOYSA-N
Synonym 1,4,8,11-tetrakis ethoxycarbonylmethyl-1,4,8,11-tetraazacyclotetradecane, tetaet, tetraethyl 1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate, tetraethyl 2,2',2,2'-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrayl tetraacetate, acmc-1c3a0, cyclam-n,n',n',n-tetraacetic acid tetraethyl ester, 1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetic acid tetraethyl ester, 1,4,8,11-tetrakis ethoxycarbonyl methyl-1,4,8,11-tetraazacyclotetradecane, 1,4,8,11-tetraazacyclotetradecane-n,n',n',n'-tetraacetic acid, tetraethyl ester, min, ethyl 2-4,8,11-tris 2-ethoxy-2-oxoethyl-1,4,8,11-tetraazacyclotetradecan-1-yl acetate
PubChem CID 14434738
IUPAC Name ethyl 2-[4,8,11-tris(2-ethoxy-2-oxoethyl)-1,4,8,11-tetrazacyclotetradec-1-yl]acetate
SMILES CCOC(=O)CN1CCCN(CC(=O)OCC)CCN(CC(=O)OCC)CCCN(CC(=O)OCC)CC1

Specifications

Melting Point 87°C to 89°C
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 544.69
Chemical Name or Material 1,4,8,11-Tetrakis(ethoxycarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.