accessibility menu, dialog, popup

UserName

CAS RN 1240257-05-9

SFFFFFClBr

CAS RN 1240257-05-9

IUPAC Name: 1-(bromomethyl)-3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-(bromomethyl)-3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 331.52
Molecular Formula: C7H5BrClF5S
InChi Key: KMRNMUYUJBXJOX-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC(Cl)=CC(CBr)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results