missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15448865
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448865 250 mg 250mg
15447217 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448865 Supplier Thermo Scientific Alfa Aesar Supplier No. H34283.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1240257-05-9
Molecular Formula C7H5BrClF5S
Molecular Weight (g/mol) 331.523
MDL Number MFCD16652423
InChI Key KMRNMUYUJBXJOX-UHFFFAOYSA-N
Synonym 3-chloro-5-pentafluorosulfur benzyl bromide, 1-bromomethyl-3-chloro-5-pentafluoro-??-sulfanyl benzene, 3-chloro-5-pentafluorothio benzyl bromide
PubChem CID 66523464
IUPAC Name [3-(bromomethyl)-5-chlorophenyl]-pentafluoro-$l^{6}-sulfane
SMILES C1=C(C=C(C=C1S(F)(F)(F)(F)F)Cl)CBr

Spezifikation

Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 331.53
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(pentafluorothio)benzyl bromide
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.