CAS RN 1227694-78-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Zaleplon Formamide, TRC
CAS: 1227694-78-1 Molecular Formula: C16H13N5O Molecular Weight (g/mol): 291.31 Synonym: N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-ethyl-formamide,Zaleplon Impurity IUPAC Name: N-[3-(3-cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-ethylformamide SMILES: CCN(C=O)c1cccc(c1)c2ccnc3c(cnn23)C#N