Learn More
Zaleplon Formamide, TRC
Product Code. 30456276
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456276 500 mg 500mg
30454467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30456276 Supplier Toronto Research Chemicals Supplier No. TRCZ145010500MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1227694-78-1
Molecular Formula C16H13N5O
Molecular Weight (g/mol) 291.31
Synonym N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-ethyl-formamide, Zaleplon Impurity
IUPAC Name N-[3-(3-cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-ethylformamide
SMILES CCN(C=O)c1cccc(c1)c2ccnc3c(cnn23)C#N
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C16H13N5O/c1-2-20(11-22)14-5-3-4-12(8-14)15-6-7-18-16-13(9-17)10-19-21(15)16/h3-8,10-11H,2H2,1H3
Formula Weight 291.112
Chemical Name or Material Zaleplon Formamide