CAS RN 117756-22-6
IUPAC Name:
disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
Synonyms:
disodium [5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl [3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxyphosphonate
Molecular Weight (g/mol):
610.27
Molecular Formula:
C15H22N2Na2O17P2
InChi Key:
PKJQEQVCYGYYMM-UHFFFAOYNA-L
SMILES:
[Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O
1
–
1
of
1
results
1
Uridine-5'-diphosphoglucose, disodium salt, 95%, MP Biomedicals™
CAS: 117756-22-6 Molecular Formula: C15H22N2Na2O17P2 Molecular Weight (g/mol): 610.27 MDL Number: MFCD00216890 InChI Key: PKJQEQVCYGYYMM-UHFFFAOYNA-L Synonym: uridine 5'-diphosphoglucose disodium,disodium 2s,3r,4s,5s-5-2,4-dioxo-3h-pyrimidin-1-yl-3,4-dihydroxyoxolan-2-yl methyl 2s,3s,4r,5r,6s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl phosphonato oxyphosphonate PubChem CID: 56846105 IUPAC Name: disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate SMILES: [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O