missing translation for 'onlineSavingsMsg'
Learn More

Uridine-5'-diphosphoglucose, disodium salt, 95%, MP Biomedicals™

Product Code. 11348829 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
500mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348829 5 g 5g
11308829 100 mg 100mg
11318829 250 mg 250mg
11328829 500 mg 500mg
11338829 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11348829 Supplier MP Biomedicals Supplier No. 0210120894
  • ONLINE EXCLUSIVE PRICE
    1960.00 EUR
    1862.70 EUR / 5g
    Save 97.30 EUR
    4% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 117756-22-6
Molecular Formula C15H22N2Na2O17P2
Molecular Weight (g/mol) 610.27
MDL Number MFCD00216890
InChI Key PKJQEQVCYGYYMM-UHFFFAOYNA-L
Synonym uridine 5'-diphosphoglucose disodium, disodium 2s,3r,4s,5s-5-2,4-dioxo-3h-pyrimidin-1-yl-3,4-dihydroxyoxolan-2-yl methyl 2s,3s,4r,5r,6s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl phosphonato oxyphosphonate
PubChem CID 56846105
IUPAC Name disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
SMILES [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O

Specifications

Color White
Assay Percent Range 95%
Quantity 5 g
Formula Weight 610.3
Physical Form Crystalline Powder
Chemical Name or Material Uridine-5'-diphosphoglucose, disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.