accessibility menu, dialog, popup

UserName

SOOOHClH2OH2O

CAS RN 1171630-97-9

IUPAC Name: 8-chloronaphthalene-1-sulfonic acid dihydrate
Synonyms: 8-chloronaphthalene-1-sulfonic acid dihydrate
Molecular Weight (g/mol): 278.7
Molecular Formula: C10H11ClO5S
InChi Key: PILNBXJFSKOZSD-UHFFFAOYSA-N
SMILES: O.O.OS(=O)(=O)C1=C2C(Cl)=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results