missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 8-Chloronaphthalene-1-sulfonic acid dihydrate, 97%

Product Code. 11450007
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450007 10 g 10g
11440007 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450007 Supplier Alfa Aesar™ Supplier No. B20948.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1171630-97-9
Molecular Formula C10H11ClO5S
Molecular Weight (g/mol) 278.70
MDL Number MFCD00052379,MFCD08533242
InChI Key PILNBXJFSKOZSD-UHFFFAOYSA-N
Synonym 8-chloronaphthalene-1-sulfonic acid dihydrate, 8-chloronaphthalene-1-sulfonic acid 2-hydrate, 8-chloronaphthalene-1-sulfonic acid-water 1/2, 8-chloronaphthalenesulfonic acid, oxamethane, oxamethane
PubChem CID 43835312
IUPAC Name 8-chloronaphthalene-1-sulfonic acid;dihydrate
SMILES O.O.OS(=O)(=O)C1=C2C(Cl)=CC=CC2=CC=C1

Specifications

Melting Point 145°C to 147°C
Quantity 10 g
UN Number UN2585
Formula Weight 278.71 (242.69 Anhydrous)
Percent Purity 97%
Chemical Name or Material 8-Chloronaphthalene-1-sulfonic acid dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.