accessibility menu, dialog, popup

UserName

CAS RN 111406-87-2

H3CNOHH2NOS

CAS RN 111406-87-2

IUPAC Name: 1-[1-(1-benzothiophen-2-yl)ethyl]-1-hydroxyurea
Synonyms: zileuton zyflo CR zyflo (TN) leutrol zyflo
Molecular Weight (g/mol): 236.29
Molecular Formula: C11H12N2O2S
InChi Key: MWLSOWXNZPKENC-UHFFFAOYNA-N
SMILES: CC(N(O)C(N)=O)C1=CC2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results