accessibility menu, dialog, popup

UserName

H3COFOOH

CAS RN 106428-05-1

IUPAC Name: 3-fluoro-2-methoxybenzoic acid
Synonyms: 3-fluoro-2-methoxybenzoic acid
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: MEOOXZGGYVXUSG-UHFFFAOYSA-N
SMILES: COC1=C(C=CC=C1F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results