missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-2-methoxybenzoic acid, 98%

Product Code. 11386539
Change view
Click to view available options
Quantity:
500 mg
2 g
Unit Size:
2g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386539 2 g 2g
11376539 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386539 Supplier Alfa Aesar™ Supplier No. H26164.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 106428-05-1
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00973972
InChI Key MEOOXZGGYVXUSG-UHFFFAOYSA-N
Synonym 3-fluoro-2-methoxybenzoicacid, 3-fluoro-2-methoxy-benzoic acid, fluoro-methoxybenzoic acid, acmc-1bo0c, buttpark 20\01-60, benzoic acid,3-fluoro-2-methoxy, 3-fluoro-2-methyloxy benzoic acid, benzoic acid, 3-fluoro-2-methoxy
PubChem CID 2737361
IUPAC Name 3-fluoro-2-methoxybenzoic acid
SMILES COC1=C(C=CC=C1F)C(=O)O

Specifications

Melting Point 112°C to 114°C
Quantity 2 g
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material 3-Fluoro-2-methoxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.