accessibility menu, dialog, popup

UserName

H3COONH2HCl

CAS RN 103565-40-8

IUPAC Name: methyl 3-[4-(aminomethyl)phenyl]propanoate hydrochloride
Synonyms: methyl 3-[4-(aminomethyl)phenyl]propanoate hydrochloride
Molecular Weight (g/mol): 229.7
Molecular Formula: C11H16ClNO2
InChi Key: DPQPQVOUGGBIGS-UHFFFAOYSA-N
SMILES: Cl.COC(=O)CCC1=CC=C(CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results