missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-[4-(aminomethyl)phenyl]propionate hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 15439115
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439115 250 mg 250mg
15429115 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439115 Supplier Thermo Scientific Alfa Aesar Supplier No. H51031.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103565-40-8
Molecular Formula C11H16ClNO2
Molecular Weight (g/mol) 229.704
MDL Number MFCD11113035
InChI Key DPQPQVOUGGBIGS-UHFFFAOYSA-N
Synonym methyl 3-4-aminomethyl phenyl propanoate hydrochloride, methyl 3-4-aminomethyl phenyl propionate hydrochloride, 3-4-aminomethyl-phenyl-propionic acid methyl ester hydrochloride, benzenepropanoic acid, 4-aminomethyl-, methyl ester, hydrochloride, benzenepropanoicacid,4-aminomethyl-,methylester,hydrochloride 1:1, acmc-20f73b, methyl 3-4-aminomethylphenyl propanoate hcl, methyl 3-4-aminomethylphenyl propionate hydrochloride, 3-4-aminomethyl-phenyl-propionic acid methyl ester hcl
PubChem CID 22282732
IUPAC Name methyl 3-[4-(aminomethyl)phenyl]propanoate;hydrochloride
SMILES COC(=O)CCC1=CC=C(C=C1)CN.Cl

Specifications

Quantity 250 mg
Beilstein 14326402
Formula Weight 229.71
Percent Purity 97%
Chemical Name or Material Methyl 3-[4-(aminomethyl)phenyl]propionate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.