accessibility menu, dialog, popup

UserName

CH3SNH2F

CAS RN 1016775-44-2

IUPAC Name: 4-fluoro-3-methylbenzene-1-carbothioamide
Synonyms: 4-fluoro-3-methylbenzenecarbothioamide
Molecular Weight (g/mol): 169.22
Molecular Formula: C8H8FNS
InChi Key: GTDSYOIDNUFACN-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results