missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF821 Polyclonal specifically detects ZNF821 in Human/Mouse samples. It is validated for ELISA/Immunocytochemistry/Immunofluorescence/Western Blot.
Specifications
Specifications
| Antigen | ZNF821 |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ16350, zinc finger protein 821 |
| Host Species | Rabbit |
| Immunogen | ZNF821 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human ZNF821. The immunogen is located within amino acids 270 - 320 of ZNF821. Amino Acid Squence: AKKSRRDNETPEEREVRR |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?