missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, TRC

Product Code. 30009850
Change view
Click to view available options
Quantity:
15 g
50 g
100 g
Unit Size:
100g
15g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009850 15 g 15g
30003383 100 g 100g
30008378 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009850 Supplier Toronto Research Chemicals Supplier No. TRCZ43890015G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2 Zn
Molecular Weight (g/mol) 136.315
Synonym Zinc chloride, Zinc chloride (ZnCl2), Zinc chloride (6CI,7CI,8CI), Butter of zinc, Knittex ZH, NSC 529648, Z 0014, Zinc Butter, Zinc dichloride, Zinc(II) chloride
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Quantity 15 g
InChI Formula InChI=1S/2ClH.Zn/h2*1H;/q;;+2/p-2
Formula Weight 133.8669
Chemical Name or Material Zinc Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.