CAS RN 7646-85-7
Zinc chloride, anhydrous, 98+%
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.28 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, 2M solution in 2-MeTHF, AcroSeal™
CAS: 7646-85-7 | Cl2Zn | 136.29 g/mol
Zinc chloride, 98.5%, for analysis
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.29 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, 98%, extra pure
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.29 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, anhydrous, 99.95% (metals basis)
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.28 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, 97+%, ACS reagent
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.29 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, 1.0M solution in diethyl ether, AcroSeal™
CAS: 7646-85-7 | Cl2Zn | 136.28 g/mol
Zinc chloride, 0.7M soln. in THF
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.28 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, ACS, 97% min
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.28 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, 1M in diethyl ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.28 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, ultra dry, 99.995% (metals basis)
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.28 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, ultra dry, 99.999% (metals basis)
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.28 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, 1M in diethyl ether, Thermo Scientific Chemicals
CAS: 7646-85-7 Molecular Formula: Cl2Zn Molecular Weight (g/mol): 136.28 MDL Number: MFCD00011295 InChI Key: JIAARYAFYJHUJI-UHFFFAOYSA-L Synonym: zinc chloride,zinc dichloride,zinc chloride zncl2,zinc butter,zinc chloride fume,zinc ii chloride,zinkchloride,zintrace,zinc chloride, anhydrous,zine dichloride PubChem CID: 5727 ChEBI: CHEBI:49976 IUPAC Name: dichlorozinc SMILES: Cl[Zn]Cl
Zinc chloride, 0.7M solution in THF, AcroSeal™
CAS: 7646-85-7 | Cl2Zn | 136.3 g/mol