missing translation for 'onlineSavingsMsg'
Learn More

(Z)-1,1,1,4,4,4-Hexafluoro-2-butene, TRC

Product Code. 30137278
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137278 5 g 5g
30136521 10 g 10g
17997444 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 30137278 Supplier Toronto Research Chemicals Supplier No. TRCH2911185G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 692-49-9
Molecular Formula C4H2F6
Molecular Weight (g/mol) 164.05
Synonym (Z)-HFO 1336, 1336mzzZ, FEA 1100, Formacel 1100, Formacel FEA 1100, HFC 1336mzz, HFO 1336mzz-Z, cis-1,1,1,4,4,4-Hexafluoro-2-butene
IUPAC Name (Z)-1,1,1,4,4,4-hexafluorobut-2-ene
SMILES FC(F)(F)\C=C/C(F)(F)F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H2F6/c5-3(6,7)1-2-4(8,9)10/h1-2H/b2-1-
Formula Weight 164.0061
Chemical Name or Material (Z)-1,1,1,4,4,4-Hexafluoro-2-butene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.