CAS RN 692-49-9
CAS RN 692-49-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(Z)-1,1,1,4,4,4-Hexafluoro-2-butene, TRC
CAS: 692-49-9 Molecular Formula: C4H2F6 Molecular Weight (g/mol): 164.05 Synonym: (Z)-HFO 1336,1336mzzZ,FEA 1100,Formacel 1100,Formacel FEA 1100,HFC 1336mzz,HFO 1336mzz-Z,cis-1,1,1,4,4,4-Hexafluoro-2-butene IUPAC Name: (Z)-1,1,1,4,4,4-hexafluorobut-2-ene SMILES: FC(F)(F)\C=C/C(F)(F)F