missing translation for 'onlineSavingsMsg'
Learn More

Xylazine, TRC

Product Code. 18073010
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18073010 1 g 1g
18042820 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18073010 Supplier Toronto Research Chemicals Supplier No. TRCX7480051G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7361-61-7
Molecular Formula C12 H16 N2 S
Molecular Weight (g/mol) 220.33
Synonym 4H-1,3-Thiazin-2-amine, N-(2,6-dimethylphenyl)-5,6-dihydro-, 4H-1,3-Thiazine, 5,6-dihydro-2-(2,6-xylidino)- (7CI,8CI), N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine, 2-(2,6-Dimethylphenylamino)-5,6-dihydro-4H-1,3-thiazine, Anased, BAY 1470, Cervizine 300, Primazine, Rometar, Sedomin, Wh 7286, Xylaz, Xylazil, Xylazin, Xylazine
IUPAC Name N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine
SMILES Cc1cccc(C)c1NC2=NCCCS2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H16N2S/c1-9-5-3-6-10(2)11(9)14-12-13-7-4-8-15-12/h3,5-6H,4,7-8H2,1-2H3,(H,13,14)
Formula Weight 220.1034
Percent Purity >95%
Chemical Name or Material Xylazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.