accessibility menu, dialog, popup

UserName

CAS RN 7361-61-7

CH3H3CHNNS

CAS RN 7361-61-7

IUPAC Name: N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine
Synonyms: xylazine
Molecular Weight (g/mol): 220.33
Molecular Formula: C12H16N2S
InChi Key: BPICBUSOMSTKRF-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1NC1=NCCCS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results