missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ XY 018

Product Code. 18792458
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792458 50 mg 50mg
18773998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18792458 Supplier R&D Systems™ Supplier No. 6223/50
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. ROR/NR1F Antagonists

XY 018 is a RORγ antagonist (EC50 = 190 nM). Suppresses androgen receptor expression in prostate cancer cell lines. Also inhibits growth of tumors in androgen receptor-positive xenograft models.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1873358-87-2
Synonym N-[2′-Fluoro-4′-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl][1,1′biphenyl]-4-yl]-2-nitrobenzeneacetamide
Quantity 50 mg
Product Line Tocris
Molecular Formula C23H15F7N2O4
Formula Weight 516.36
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.