missing translation for 'onlineSavingsMsg'
Learn More

Wnt-2b Antibody, Novus Biologicals™

Product Code. 18214815 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214815 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214815 Supplier Novus Biologicals Supplier No. NBP153123
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Wnt-2b Polyclonal specifically detects Wnt-2b in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Wnt-2b
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q93097
Gene Alias Protein Wnt-13, protein Wnt-2b, wingless-type MMTV integration site family, member 2B, WNT13XWNT2, Xenopus, homolog of, XWNT2wingless-type MMTV integration site family, member 13
Gene Symbols WNT2B
Host Species Rabbit
Immunogen Synthetic peptides corresponding to WNT2B(wingless-type MMTV integration site family, member 2B) The peptide sequence was selected from the N terminal of WNT2B. Peptide sequence MLRPGGAEEAAQLPLRRASAPVPVPSPAAPDGSRASARLGLACLLLLLLL.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7482
Test Specificity Expected identity based on immunogen sequence: Human: 100%; Rabbit: 91%; Pig: 85%; Bovine: 78%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.