missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Wnt-1 Monoclonal specifically detects Wnt-1 in Human, Mouse samples. It is validated for ELISA, Western Blot.
Specifications
Specifications
| Antigen | Wnt-1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 13F9 |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | INT1Proto-oncogene Int-1 homolog, proto-oncogene Wnt-1, wingless-type MMTV integration site family, member 1, wingless-type MMTV integration site family, member 1 (oncogene INT1) |
| Host Species | Mouse |
| Immunogen | Synthetic peptide corresponding to an internal region of human Wnt-1 protein. (Uniprot: P04628) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?