missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VIP Monoclonal specifically detects VIP in Bovine, Human, Pig, Rat samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications
Specifications
| Antigen | VIP |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 2 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 |
| Formulation | 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide |
| Gene Accession No. | P01282 |
| Gene Alias | MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?