Specifications
| Content And Storage | Store at 4°C. Do not freeze. |
| For Use With (Application) | Western Blot, ELISA, Immunohistochemistry (IHC), Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen), ISH, ELISpot |
| Species | Non-species specific |
| Quantity | 1 Kit |