Learn More
Vanillyl Methyl Ketone, TRC
Product Code. 30450234
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450234 1 g -
30455068 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30450234 Supplier Toronto Research Chemicals Supplier No. TRCV0976251G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2503-46-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
Synonym 1-(4-Hydroxy-3-methoxyphenyl)-2-propanone, (4-Hydroxy-3-methoxyphenyl)acetone, 1-(4-Hydroxy-3-methoxyphenyl)-2-propanone, 2-Propiovanillone, Guaiacylacetone, Methyl Vanillyl Ketone, NSC 16690
IUPAC Name 1-(4-hydroxy-3-methoxyphenyl)propan-2-one
SMILES COc1cc(CC(=O)C)ccc1O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C10H12O3/c1-7(11)5-8-3-4-9(12)10(6-8)13-2/h3-4,6,12H,5H2,1-2H3
Formula Weight 180.0786
Chemical Name or Material Vanillyl Methyl Ketone