CAS RN 2503-46-0
IUPAC Name:
1-(4-hydroxy-3-methoxyphenyl)propan-2-one
Synonyms:
1-(4-hydroxy-3-methoxyphenyl)propan-2-one
Molecular Weight (g/mol):
180.2
Molecular Formula:
C10H12O3
InChi Key:
LFVCJQWZGDLHSD-UHFFFAOYSA-N
SMILES:
COC1=CC(CC(C)=O)=CC=C1O
1
–
1
of
1
results
1
Vanillyl Methyl Ketone, TRC
CAS: 2503-46-0 Molecular Formula: C10H12O3 Molecular Weight (g/mol): 180.2 Synonym: 1-(4-Hydroxy-3-methoxyphenyl)-2-propanone,(4-Hydroxy-3-methoxyphenyl)acetone,1-(4-Hydroxy-3-methoxyphenyl)-2-propanone,2-Propiovanillone,Guaiacylacetone,Methyl Vanillyl Ketone,NSC 16690 IUPAC Name: 1-(4-hydroxy-3-methoxyphenyl)propan-2-one SMILES: COc1cc(CC(=O)C)ccc1O