missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VAChT/SLC18A3 Monoclonal specifically detects VAChT/SLC18A3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry.
Specifications
Specifications
| Antigen | VAChT/SLC18A3 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | S6-38 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter |
| Gene Symbols | SLC18A3 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?