missing translation for 'onlineSavingsMsg'
Learn More

UbcH8/Ube2L6 Antibody (k1H3), DyLight 755, Novus Biologicals™

Product Code. 18214863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214863 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214863 Supplier Novus Biologicals Supplier No. NBP104351IR
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

UbcH8/Ube2L6 Monoclonal specifically detects UbcH8/Ube2L6 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UbcH8/Ube2L6
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone k1H3
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 6.3.2.19, retinoic acid induced gene B protein, Retinoic acid-induced gene B protein, RIG-B, UbcH8, UBCH8MGC40331, Ubiquitin carrier protein L6, ubiquitin/ISG15-conjugating enzyme E2 L6, ubiquitin-conjugating enzyme E2L 6, Ubiquitin-protein ligase L6
Gene Symbols UBE2L6
Host Species Mouse
Immunogen Recombinant human UBE2L6 (1-152aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9246
Test Specificity Anti-human UBE2L6 mAb, clone k1H3, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human UBE2L6 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.