missing translation for 'onlineSavingsMsg'
Learn More

TYW5 Antibody, Novus Biologicals™

Product Code. 18275479 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18275479 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18275479 Supplier Novus Biologicals Supplier No. NBP191415

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TYW5 Polyclonal specifically detects TYW5 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TYW5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_001034782
Gene Alias tRNA-yW synthesizing protein 5
Gene Symbols TYW5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C2orf60. Peptide sequence HLPSECYDKTDTYGNKDPTAASRAAQILDRALKTLAELPEEYRDFYARRM.
Molecular Weight of Antigen 36 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 129450
Test Specificity Expected identity based on immunogen sequence: Bacillus subtilis subsp. natto BEST195: 90%; Bacillus subtilis: 90%; Rat: 85%; Black-legged tick: 83%; Zebrafish: 78%; Green puffer: 78%; Rat: 86%; Mouse: 86%; Zebrafish: 79%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.