Learn More
Tris(tetrabutylammonium) Hydrogen Pyrophosphate, TRC
Product Code. 30456669
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456669 5 g 5g
30454559 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30456669 Supplier Toronto Research Chemicals Supplier No. TRCT8846005G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 76947-02-9
Molecular Formula C48H109N3O7P2
Molecular Weight (g/mol) 902.34
Synonym Tris(tetrabutylammonium) Pyrophosphate, Tris(N,N,N-tributyl-1-butanaminium) Diphosphate
IUPAC Name [hydroxy(oxido)phosphoryl] phosphate;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.OP(=O)([O-])OP(=O)([O-])[O-]
Spezifikation
Quantity 5 g
InChI Formula InChI=1S/3C16H36N.H4O7P2/c3*1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-8(2,3)7-9(4,5)6/h3*5-16H2,1-4H3;(H2,1,2,3)(H2,4,5,6)/q3*+1;/p-3
Formula Weight 901.7741
Chemical Name or Material Tris(tetrabutylammonium) Hydrogen Pyrophosphate