CAS RN 76947-02-9
CAS RN 76947-02-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tris(tetrabutylammonium) Hydrogen Pyrophosphate, TRC
CAS: 76947-02-9 Molecular Formula: C48H109N3O7P2 Molecular Weight (g/mol): 902.34 Synonym: Tris(tetrabutylammonium) Pyrophosphate,Tris(N,N,N-tributyl-1-butanaminium) Diphosphate IUPAC Name: [hydroxy(oxido)phosphoryl] phosphate;tetrabutylazanium SMILES: CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.OP(=O)([O-])OP(=O)([O-])[O-]