Learn More
Trimethylamine Monohydrochloride, TRC
Product Code. 30010006
Change view
Click to view available options
Quantity:
500 mg
5 g
10 g
Unit Size:
10g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010006 500 mg 500mg
30005873 10 g 10g
30006782 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010006 Supplier Toronto Research Chemicals Supplier No. TRCT796968500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 593-81-7
Molecular Formula C3 H9 N . Cl H
Molecular Weight (g/mol) 95.5712
Synonym Hegzadesil, Trimethylamine Hydrochloric Acid, Trimethylammonium Chloride, N,N-Dimethylmethanamine Hydrochloride
IUPAC Name N,N-dimethylmethanamine;hydrochloride
SMILES Cl.CN(C)C
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C3H9N.ClH/c1-4(2)3;/h1-3H3;1H
Formula Weight 95.0502
Chemical Name or Material Trimethylamine Monohydrochloride