accessibility menu, dialog, popup

UserName

CAS RN 593-81-7

HClCH3NCH3H3C

CAS RN 593-81-7

IUPAC Name: hydrogen trimethylamine chloride
Synonyms: hydrogen trimethylamine chloride
Molecular Weight (g/mol): 95.57
Molecular Formula: C3H10ClN
InChi Key: SZYJELPVAFJOGJ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)