Learn More
Novus Biologicals™ TRIM50C 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| Research Category | Zinc Finger |
| Target | TRIM50C |
| Synonym | MGC45440, TRIM50CTripartite motif-containing protein 50 C, tripartite motif containing 74, tripartite motif-containing 50 C, tripartite motif-containing 74, tripartite motif-containing protein 74 |
| Gene Symbol | TRIM74 |